Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfosuccinic acid solution

Catalog No. 3385671L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-856-71L 1 L
33-856-7250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-856-71L Supplier Sigma Aldrich Supplier No. 3385671L
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sulfosuccinic acid solution

Specifications

CAS 5138-18-1
Density 1.438 g/mL (at 25°C)
Molecular Formula C4H6O7S
Refractive Index n20/D 1.449
Linear Formula HOOCCH2CH(SO3H)COOH
Quantity 1 L
MDL Number MFCD00075139
Synonym 2-Sulfobutanedioic acid
Molecular Weight (g/mol) 198.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.