Learn More
Cayman Chemical Sulfosuccinimidyl 3-[[2-(Biotinamido)ethyl]dithio]propionate (sodium salt), CAS: 325143-98-4, 100 mg
SDP
Catalog No. 502917327
Encompass_Preferred
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii
Catalog No. 50-291-7327 Supplier Cayman Chemical Supplier No. 39537100 MG
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii