Learn More
Cayman Chemical Sulfosuccinimidyl 3-[[2-(Biotinamido)ethyl]dithio]propionate (sodium salt), CAS: 325143-98-4, 25 mg
SDP
Catalog No. 502916981
Encompass_Preferred
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii
Catalog No. 50-291-6981 Supplier Cayman Chemical Supplier No. 3953725 MG
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii