Learn More
Cayman Chemical SulfosuccInImIdyl 32 BIot 50mg
SDP
Catalog No. 502916982
Encompass_Preferred
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii
Catalog No. 50-291-6982 Supplier Cayman Chemical Supplier No. 3953750 MG
An amine-reactive reagent; has been used as a derivatization reagent to identify PTMs in lysine-rich proteins and novel cell surface proteins of R. rickettsii