Learn More
Cayman Chemical Sulfosuccinimidyl Myristate (sodium salt), Purity: ≥95%, CAS: 162823-01-0, 5 mg
SDP
Catalog No. 502951532
Encompass_Preferred
An inhibitor of fatty acid transport that has been used as an affinity label for certain membrane proteins that bind to fatty acids
Catalog No. 50-295-1532 Supplier Cayman Chemical Supplier No. 112125 MG
An inhibitor of fatty acid transport that has been used as an affinity label for certain membrane proteins that bind to fatty acids