Learn More
Medchemexpress LLC HY-112103 100mg Medchemexpress, 6-(γ,γ-Dimethylallylamino)purine CAS:2365-40-4 Purity:>98%
SDP
Catalog No. 501969024
Encompass_Preferred
Medchemexpress, HY-112847A 10mg Sulfosuccinimidyl oleate (sodium) CAS: Sulfosuccinimidyl oleate sodium (Sulfo-N-succinimidyl oleate sodium) is a long chain fatty acid that inhibits fatty acid transport into cells. Sulfosuccinimidyl oleate sodium is a potent and irreversible inhibitor of mitochondrial respiratory chain. Sulfosuccinimidyl oleate sodium binds the CD36 receptor on the surface of microglia. Anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9024 Supplier Medchemexpress LLC Supplier No. HY112103100MG
Medchemexpress, HY-112847A 10mg Sulfosuccinimidyl oleate (sodium) CAS: Sulfosuccinimidyl oleate sodium (Sulfo-N-succinimidyl oleate sodium) is a long chain fatty acid that inhibits fatty acid transport into cells. Sulfosuccinimidyl oleate sodium is a potent and irreversible inhibitor of mitochondrial respiratory chain. Sulfosuccinimidyl oleate sodium binds the CD36 receptor on the surface of microglia. Anti-inflammatory effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.