Learn More
TARGETMOL CHEMICALS INC SULFOSUCCINIMIDYL OLEATE 10MG
SDP
Catalog No. 502577405
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Sulfosuccinimidyl oleate sodium (Sulfo-N-succinimidyl oleate sodium) is a long-chain fatty acid that inhibits the transport of fatty acids to cells. Sulfosuccinimidyl oleate sodium is an effective irreversible inhibitor of mitochondrial respiratory chain. Sulfosuccinimidyl oleate sodium binds to CD36 receptors on the surface of microglia. purity: 98%
Catalog No. 50-257-7405 Supplier TARGETMOL CHEMICALS INC Supplier No. T13036L10MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Sulfosuccinimidyl oleate sodium (Sulfo-N-succinimidyl oleate sodium) is a long-chain fatty acid that inhibits the transport of fatty acids to cells. Sulfosuccinimidyl oleate sodium is an effective irreversible inhibitor of mitochondrial respiratory chain. Sulfosuccinimidyl oleate sodium binds to CD36 receptors on the surface of microglia. purity: 98%