Learn More
Medchemexpress LLC HY-112847A 5mg Medchemexpress, Sulfosuccinimidyl oleate sodium CAS: Purity:>98%
SDP
Catalog No. 501932442
Encompass_Preferred
Medchemexpress, HY-112847A 5mg Sulfosuccinimidyl oleate sodium CAS: Sulfosuccinimidyl oleate sodium is a long chain fatty acid that inhibits fatty acid transport into cells. Sulfosuccinimidyl oleate binds the CD36 receptor on the surface of microglia. Anti-inflammatory effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2442 Supplier Medchemexpress LLC Supplier No. HY112847A5MG
Medchemexpress, HY-112847A 5mg Sulfosuccinimidyl oleate sodium CAS: Sulfosuccinimidyl oleate sodium is a long chain fatty acid that inhibits fatty acid transport into cells. Sulfosuccinimidyl oleate binds the CD36 receptor on the surface of microglia. Anti-inflammatory effect [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.