Learn More
Medchemexpress LLC Sulfosuccinimidyl oleate (sodium) | 1212012-37-7 | 99.7% | 481.58 | 50 MG
SDP
Catalog No. 5000437756
Encompass_Preferred
Sulfosuccinimidyl oleate sodium is a long-chain fatty acid that inhibits fatty acid transport into cells. It is a potent and irreversible inhibitor of the mitochondrial respiratory chain and binds to the CD36 receptor on the surface of microglia, demonstrating an anti-inflammatory effect.
  • Inhibits fatty acid transport into cells.
  • Potent and irreversible inhibitor of mitochondrial respiratory chain.
  • Binds the CD36 receptor on the surface of microglia.
  • Exhibits anti-inflammatory effect.
  • Neuroprotective and alleviates stroke-induced neuroinflammation.
Catalog No. 50-004-37756 Supplier Medchemexpress LLC Supplier No. HY112847A50MG
Sulfosuccinimidyl oleate sodium is a long-chain fatty acid that inhibits fatty acid transport into cells. It is a potent and irreversible inhibitor of the mitochondrial respiratory chain and binds to the CD36 receptor on the surface of microglia, demonstrating an anti-inflammatory effect.
  • Inhibits fatty acid transport into cells.
  • Potent and irreversible inhibitor of mitochondrial respiratory chain.
  • Binds the CD36 receptor on the surface of microglia.
  • Exhibits anti-inflammatory effect.
  • Neuroprotective and alleviates stroke-induced neuroinflammation.