Learn More
Cayman Chemical Sulfosuccinimidyl Oleate (sodium salt), Purity: ≥95%, CAS: 1212012-37-7, 5 mg
SDP
Catalog No. 502951077
Encompass_Preferred
An irreversible inhibitor of the fatty acid translocase CD36, blocking uptake of oleate, linoleate, or stearate by about 65% when added at 200 µM to adipocytes; reduces the uptake of palmitate by mouse insulinoma MIN6 cells and RAW 264.7 macrophages
Catalog No. 50-295-1077 Supplier Cayman Chemical Supplier No. 112115 MG
An irreversible inhibitor of the fatty acid translocase CD36, blocking uptake of oleate, linoleate, or stearate by about 65% when added at 200 µM to adipocytes; reduces the uptake of palmitate by mouse insulinoma MIN6 cells and RAW 264.7 macrophages