Learn More
Bachem H-Tyr(SO3H)-OH
SDP
Supplier:  Bachem 4033889.0001
Encompass_Preferred
H-Tyr(SO3H)-OH Sodium, 956-46-7, C9H11NO6S, Mr 261.26, 1g. Sulfotyrosine has been used by Liu et al. for conveniently obtaining tyrosine-sulfated proteins by expression in E. coli with an expanded genetic code.
Catalog No. 50-194-7360