Learn More
Hach Company Sulfuric Acid Standardette Solution #100, Pack of 12
Catalog No. NC2940726 Shop All Hach Company Products
Encompass_Preferred
The Sulfuric Acid Standardette Solution #100 was designed to ensure precision, efficiency and standardization of 0.100 N base titrants when performing titrations in your lab. With a unique method of standardizing solutions, these single use bottles will make your process faster, easier and more accurate than traditional methods.
Catalog No. NC2940726 Supplier Hach Company Supplier No. ATS100
The Sulfuric Acid Standardette Solution #100 was designed to ensure precision, efficiency and standardization of 0.100 N base titrants when performing titrations in your lab. With a unique method of standardizing solutions, these single use bottles will make your process faster, easier and more accurate than traditional methods.