Learn More
Revvity Health Sciences Inc Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Catalog No. 502810966
Encompass
Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water
Catalog No. 50-281-0966 Supplier Revvity Health Sciences Inc Supplier No. NEX042005MC
Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sulfuric Acid in 1mL Water