Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur in Isooctane standard solution, Specpure™, 1000μg/g (0.100%)

Catalog No. AA41324AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41324AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41324AE Supplier Thermo Scientific Chemicals Supplier No. 041324AE
Add to Cart

missing translation for 'validMainframeMsg'

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00804269

Specifications

UN Number UN1262
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Isooctane standard solution, 1000μg/g (0.100%)
Hazard Category H225-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECSNumber 208-759-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.