Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur Oil Standard in No. 2 Diesel Fuel, 10μg/g, Bulk, SPEX CertiPrep™
SDP

Catalog No. SDFSBLKY Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Triple-leached LDPE bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SDFS-BLK-Y 100 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDFS-BLK-Y Supplier SPEX CertiPrep Supplier No. SDFSBLKY
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Sulfur Oil Standard Blank in #2 Diesel Fuel, SPEX CertiPrep. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Sulfur Oil Standard Blank
  • For use in diesel fuel analysis
  • 1 mL volume in #2 Diesel Fuel

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 68476-34-6

Specifications

Physical Form Liquid
Quantity 100 mL
Packaging Triple-leached LDPE bottle
Chemical Name or Material Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis
Hazard Category Causes severe skin burns and eye damage
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.