Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfur Oil Standard in No. 2 Diesel Fuel, 100μg/g, SPEX CertiPrep™
SDP

Catalog No. SDFS100Y Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDFS-100-Y 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDFS-100-Y Supplier SPEX CertiPrep Supplier No. SDFS100Y
Add to Cart
Add to Cart

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 544-40-1 , 68476-34-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.29
MDL Number MFCD00009468
InChI Key HTIRHQRTDBPHNZ-UHFFFAOYSA-N
PubChem CID 11002
IUPAC Name 1-butylsulfanylbutane
SMILES CCCCSCCCC

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 99.99, 0.01
Analyte or Component Names Sulphur
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Element Sulphur
Solvent or Matrix #2 Diesel Fuel
Industry Type Petrochemical Testing
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 100μg/g
Chemical Name or Material Sulfur Oil Standard in #2 Diesel Fuel, For Diesel Fuel Analysis
Show More Show Less
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.