Learn More
Strem, An Ascensus Company CAS# 7704-34-9. 1kg. Sulfur powder, precipitated, purified. MFCD00085316
SDP
Catalog No. 5090114251
Encompass
CAS# 7704-34-9. 1kg. Sulfur powder, precipitated, purified. MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -60 mesh pwdr. Strem# 93-1617. http://www.strem.com/catalog/v/93-1617/
Catalog No. 50-901-14251 Supplier Strem, An Ascensus Company Supplier No. 9316171000G
CAS# 7704-34-9. 1kg. Sulfur powder, precipitated, purified. MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -60 mesh pwdr. Strem# 93-1617. http://www.strem.com/catalog/v/93-1617/