Learn More
Strem, An Ascensus Company CAS# 7704-34-9. 250g. Sulfur powder, precipitated, purified. MFCD00085316
SDP
Catalog No. 5090114250
Encompass
CAS# 7704-34-9. 250g. Sulfur powder, precipitated, purified. MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -60 mesh pwdr. Strem# 93-1617. http://www.strem.com/catalog/v/93-1617/
Catalog No. 50-901-14250 Supplier Strem, An Ascensus Company Supplier No. 931617250G
CAS# 7704-34-9. 250g. Sulfur powder, precipitated, purified. MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -60 mesh pwdr. Strem# 93-1617. http://www.strem.com/catalog/v/93-1617/