Learn More
Strem, An Ascensus Company CAS# 7704-34-9. 250g. Sulfur powder, sublimed (99+%). MFCD00085316
SDP
Catalog No. 5090114252
Encompass
CAS# 7704-34-9. 250g. Sulfur powder, sublimed (99+%). MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -100 mesh pwdr. Strem# 93-1618. http://www.strem.com/catalog/v/93-1618/
Catalog No. 50-901-14252 Supplier Strem, An Ascensus Company Supplier No. 931618250G
CAS# 7704-34-9. 250g. Sulfur powder, sublimed (99+%). MFCD00085316. Molecular Weight: 32.06. Molecular Formula: S. Color/form: -100 mesh pwdr. Strem# 93-1618. http://www.strem.com/catalog/v/93-1618/