Learn More
Strem, An Ascensus Company CAS# 201611-92-9. 2g. 4-Cyano-4-(thiobenzoylthio)pentanoic acid, min. 97%. MFCD10698690
SDP
Supplier:  Strem, An Ascensus Company 1604222G
Encompass
CAS# 201611-92-9. 2g. 4-Cyano-4-(thiobenzoylthio)pentanoic acid, min. 97%. MFCD10698690. Molecular Weight: 279.38. Molecular Formula: C13H13NO2S2. Color/form: pink pwdr. Strem# 16-0422. http://www.strem.com/catalog/v/16-0422/
Catalog No. 50-204-5265