Learn More
Perkin Elmer US LLC Sulfur (S) Standard, 100 µg/g, in Hydrocarbon Oil, 50 g
Supplier:  Perkin Elmer US LLC N9308230
Encompass_Preferred
Symbol: S Matrix: Hydrocarbon Oil Size: 50 g Features and Benefits Metallo-organic metals in hydrocarbon oil Accuracy ensured by Quality Testing with NIST Standard Reference Materials when available Certificate of Analysis supplied with each standard
Catalog No. 50-210-3806