Learn More
SPEX CERTIPREP LLC 3P SULFUR STD 1000PPM, 100ML
Catalog No. NC0659967
Encompass_Preferred
(HPS by Zeptometrix) S 1000ug/mL in H2O Exp Date: 18 Months Hazardous by air This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0659967 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000545100
(HPS by Zeptometrix) S 1000ug/mL in H2O Exp Date: 18 Months Hazardous by air This standard is traceable to NIST and certified Guide 34 and 17025.