Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfur Trioxide Trimethylamine Complex

Catalog No. 135879100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-587-9100G 100 g
13-587-95G 5 g
13-587-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-587-9100G Supplier Sigma Aldrich Supplier No. 135879100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sulfur Trioxide Trimethylamine Complex

Specifications

CAS 3162-58-1
Melting Point 232°C
Molecular Formula C3H9NO3S
Linear Formula (CH3)3N -+ SO3
MDL Number MFCD00012421
Quantity 100 g
Synonym NSC 9838; Trimethylamine sulfur trioxide complex
Molecular Weight (g/mol) 139.17 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.