Learn More
TARGETMOL CHEMICALS INC SULFURETIN 5MG
SDP
Catalog No. 502578878
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Sulfuretin is a potent anti-oxidant has protective effect against t-BHP-induced oxidative damage in human liver-derived HepG2 cells is attributable to its ability to scavenge ROS and up-regulate the activity of HO-1 through the Nrf2/ARE and JNK/ERK signaling pathways. purity: 98%
Catalog No. 50-257-8878 Supplier TARGETMOL CHEMICALS INC Supplier No. TN22445MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Sulfuretin is a potent anti-oxidant has protective effect against t-BHP-induced oxidative damage in human liver-derived HepG2 cells is attributable to its ability to scavenge ROS and up-regulate the activity of HO-1 through the Nrf2/ARE and JNK/ERK signaling pathways. purity: 98%