Learn More
LaMarKa Chemical Company Sulfuric Acid 0.02N Phenolphthalein Endpoint, 1l
Catalog No. 504400428
Encompass_Preferred
Sulfuric Acid 0.02N Phenolphthalein Endpoint, 1l Plastic Bottle
Catalog No. 50-440-0428 Supplier LaMarKa Chemical Company Supplier No. 0000170216750101
Sulfuric Acid 0.02N Phenolphthalein Endpoint, 1l Plastic Bottle