Learn More
Hach Company Sulfuric Acid Standard Solution, 0.035 N, 100 mL MDB
Supplier:  Hach Company 2349732
Encompass
For alkalinity determination by drop-count titration test kit. 100 mL Marked Dropping Bottle
Catalog No. 50-199-5847