Learn More
Hach Company Sulfuric Acid Standard Solution, 0.100 N (N/10), 1 L
Supplier:  Hach Company 20253
Encompass
Standard acid titrating solution. For various titration procedures and pH adjustments. 0.100 N (N/10), 1 liter bottle
Catalog No. 50-199-4200