Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.1142 N, Ricca Tightrant™
SDP

Catalog No. 82555 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8255-5 20 L Cubitainer
8255-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8255-5 Supplier Ricca Chemical Company Supplier No. 82555
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for Crude Fiber Analysis of Food and Feed

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

Molecular Formula H2O4S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % 97.0, 0.57
Color Colorless
CAS 7664-93-9, 7732-18-5
Physical Form Liquid
Quantity 20 L
CAS Max % 0.6
Packaging Cubitainer
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.1142Normal
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.