Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.16 Normal Standardized, Reagents

Catalog No. 01333956
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-956 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-956 Supplier Reagents Holdings Llc Supplier No. CS0126004A
Add to Cart
Add to Cart

Sulfuric Acid, 0.16 Normal Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfuric Acid, 0.16 Normal Standardized
pH <1
Color Colorless
CAS 7732-18-5, 7664-93-9
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0038
Quantity 4 L
Molecular Formula H2SO4/H2O
Physical Form Liquid
Synonym Sulphuric Acid, Dihydrogen Sulfate
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.