Learn More
Hach Company Sulfuric Acid Digital Titrator Cartridge, 0.1600 N
Catalog No. 501995013 Shop All Hach Company Products
Encompass
For determination of alkalinity by Digital Titrator titration. Hach method 8203. 0.1600 N Sulfuric Acid. Digits = mg/L CaCO3 in 100 mL, Digits x 0.002 = meq. Approximately 40-100 tests.
This item is not returnable. View return policy
Catalog No. 50-199-5013 Supplier Hach Company Supplier No. 1438801
 Restricted Item
Add to Cart
Add to Cart
For determination of alkalinity by Digital Titrator titration. Hach method 8203. 0.1600 N Sulfuric Acid. Digits = mg/L CaCO3 in 100 mL, Digits x 0.002 = meq. Approximately 40-100 tests.