Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.2284 Normal Standardized, Reagents

Catalog No. 01333831
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-831 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-831 Supplier Reagents Holdings Llc Supplier No. CS1048004A
Only null left
Add to Cart
Add to Cart

Sulfuric Acid, 0.2284 Normal Standardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Specifications

Chemical Name or Material Sulfuric Acid, 0.2284 Normal Standardized
pH <1
Color Colorless
CAS 7732-18-5, 7664-93-9
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0075
Quantity 4 L
Molecular Formula H2SO4/H2O
Physical Form Liquid
Synonym Sulphuric Acid, Dihydrogen Sulfate
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.