Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.250 N (N/4), Ricca Tightrant™
SDP

Catalog No. R8270000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R8270000500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R8270000500 Supplier Ricca Chemical Company Supplier No. 827016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified volumetric titrant suitable for use in TAPPI crude tall oil analysis and APHA mercury testing.

  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Specifications

Color Colorless
CAS 7732-18-5, 7664-93-9
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Assay Percent Range 0.2500 ± 0.0001 N at 20°C
Quantity 500 mL
Molecular Formula H2O4S
Physical Form Liquid
MDL Number MFCD00064589
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Solubility Information Solubility in Water: Infinite
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Grade Laboratory
Chemical Name or Material Sulfuric Acid, 0.250 Normal (N/4), TP wash
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.