Learn More
Hach Company Sulfuric Acid Standard Solution, 0.500 N, 100 mL
Catalog No. 501994700 Shop All Hach Company Products
Encompass
For alkalinity determination by drop-count titration
Catalog No. 50-199-4700 Supplier Hach Company Supplier No. 212132
For alkalinity determination by drop-count titration