Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Sulfuric Acid 1 0N 20L
SDP

Catalog No. 5000550956
Encompass_Preferred

Sulfuric Acid 1 0N 20L

Catalog No. 50-005-50956 Supplier Thomas Scientific Supplier No. C748C30
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 1 0N 20L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.