Learn More
Hach Company Sulfuric Acid Digital Titrator Cartridge, 1.600 N
Supplier:  Hach Company 1438901
Encompass
For determination of alkalinity by Digital Titrator titration. Hach method 8203. 1.600 N Sulfuric Acid. Digits = mg/L CaCO3 in 100 mL, Digits x 0.002 = meq. Approximately 40-100 tests.
Catalog No. 50-199-5014