Learn More
Hach Company Sulfuric Acid Standard Solution, 19.2 N, 100 mL MDB
Supplier:  Hach Company 203832
Encompass
Multi-purpose sulfuric acid standard solution. Required for determination of volatile acids by the esterification method. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4661