Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Sulfuric Acid 2 0N 20L UN3264
SDP

Catalog No. 5000550711
Encompass_Preferred

Sulfuric Acid 2 0N 20L UN3264

Catalog No. 50-005-50711 Supplier Thomas Scientific Supplier No. C748D41
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 2 0N 20L UN3264

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.