Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Sulfuric Acid 0.2 Normal NIST Traceable, Solution (4L)
SDP

Catalog No. NC0676151
Encompass_Preferred

Sulfuric Acid 0.2 Normal NIST Traceable, Solution (4L)

Catalog No. NC0676151 Supplier Aqua Solutions Supplier No. 90504L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 0.2 Normal NIST Traceable, Solution (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.