Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Sulfuric Acid 50% (v/v) 1L UN1830
SDP

Catalog No. 5000550856
Encompass_Preferred

Sulfuric Acid 50% (v/v) 1L UN1830

Catalog No. 50-005-50856 Supplier Thomas Scientific Supplier No. C748C36
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 50% (v/v) 1L UN1830

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.