Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thomas Scientific Sulfuric Acid 6 0N 500mL UN2796
SDP

Catalog No. 5000550596
Encompass_Preferred

Sulfuric Acid 6 0N 500mL UN2796

Catalog No. 50-005-50596 Supplier Thomas Scientific Supplier No. C748D34
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid 6 0N 500mL UN2796

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.