Learn More
Hach Company BioTector B3500 Reagent, 6.0 N Sulfuric Acid with Mn Catalyst, 20 L
Catalog No. NC1776991 Shop All Hach Company Products
Encompass
6.0 N sulfuric acid reagent with a manganese catalyst supplied in a 20 L container for use with BioTector B3500 TOC analyzers. Intended for total organic carbon (TOC) analysis and instrument reagent use. Store in a cool, dry place and consult the safety data sheet before handling.
  • 6.0 N sulfuric acid formulated for TOC reagent protocols.
  • Contains manganese catalyst to support oxidation reactions.
  • Supplied in a 20 L container suitable for laboratory instrument use.
  • Compatible with online and benchtop TOC analyzers that require acidic reagent.
  • Store in a cool, dry, well-ventilated area and protect from moisture.
  • Safety data sheet available and required for safe handling.
Catalog No. NC1776991 Supplier Hach Company Supplier No. 2038162
6.0 N sulfuric acid reagent with a manganese catalyst supplied in a 20 L container for use with BioTector B3500 TOC analyzers. Intended for total organic carbon (TOC) analysis and instrument reagent use. Store in a cool, dry place and consult the safety data sheet before handling.
  • 6.0 N sulfuric acid formulated for TOC reagent protocols.
  • Contains manganese catalyst to support oxidation reactions.
  • Supplied in a 20 L container suitable for laboratory instrument use.
  • Compatible with online and benchtop TOC analyzers that require acidic reagent.
  • Store in a cool, dry, well-ventilated area and protect from moisture.
  • Safety data sheet available and required for safe handling.