Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Sulfuric acid 78% w/w | 7664-93-9 | MFCD00064589 | 1 L
SDP

Catalog No. NC1028925
Encompass

Sulfuric acid 78% w/w | Mol Wt: 98.08 | 7664-93-9 | MFCD00064589 | 1 L

Catalog No. NC1028925 Supplier Aqua Solutions Supplier No. COV001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric acid 78% w/w | Mol Wt: 98.08 | 7664-93-9 | MFCD00064589 | 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.