Learn More
Hach Company Sulfuric Acid Digital Titrator Cartridge, 8.00 N
Supplier:  Hach Company 1439101
Encompass
For base concentration determination by Digital Titrator titration. Digits x 0.01 = meq
Catalog No. 50-199-5016