Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andwin Scientific SULFURIC ACID, 96%, ACS/F 2.5L
SDP

Catalog No. NC2075306
Encompass_Preferred

Sulfuric Acid, 96%, ACS/FCC H2SO4,

Catalog No. NC2075306 Supplier Andwin Scientific Supplier No. BDH30722.5LG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid, 96%, ACS/FCC H2SO4,

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.