Learn More
Medchemexpress LLC Sulfuric acid ammonium salt (1:2) | 7783-20-2 | 99.3% | 132.14 | 50 G
SDP
Catalog No. 5000365853
Encompass_Preferred
Ammonium sulphate, ≥99.0%, AR is an inorganic sulfate salt primarily used in molecular biology. It serves to precipitate proteins, separate antibodies, and enhance antigen-antibody reactions. Beyond laboratory applications, it acts as a nitrogen source in plant fertilizers.
  • Utilized in molecular biology
  • Can precipitate proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Functions as a nitrogen source in plant fertilizers
  • Exhibits developmental toxicity to freshwater snails
  • Increases population of *Anopheles africanus* and *Culex* mosquito larvae in rice paddies
  • Inhibits conversion of glucose to ascorbic acid in plants
Catalog No. 50-003-65853 Supplier Medchemexpress LLC Supplier No. 50GHYY0261A
Ammonium sulphate, ≥99.0%, AR is an inorganic sulfate salt primarily used in molecular biology. It serves to precipitate proteins, separate antibodies, and enhance antigen-antibody reactions. Beyond laboratory applications, it acts as a nitrogen source in plant fertilizers.
  • Utilized in molecular biology
  • Can precipitate proteins
  • Separates antibodies
  • Enhances antigen-antibody reactions
  • Functions as a nitrogen source in plant fertilizers
  • Exhibits developmental toxicity to freshwater snails
  • Increases population of *Anopheles africanus* and *Culex* mosquito larvae in rice paddies
  • Inhibits conversion of glucose to ascorbic acid in plants