Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Fuming, puriss. p.a., Solstice

Catalog No. 6001631 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-016-31 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-016-31 Supplier Solstice Supplier No. 307361L

missing translation for 'validMainframeMsg'

Puriss. p.a., 20% SO3

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8014-95-7
Molecular Formula H2O7S2
Molecular Weight (g/mol) 178.13
MDL Number MFCD00137792
InChI Key HIFJUMGIHIZEPX-UHFFFAOYSA-N
Synonym oleum, fuming sulfuric acid, sulfuric acid, fuming, oleum iodisum, sulfuric acid fuming, oleum fuming sulfuric acid, sulfuric acid, mixt with sulfur trioxide, sulfur trioxide, mixt. with sulfuric acid, sulfuric acid mixture with sulfur trioxide
PubChem CID 24681
IUPAC Name sulfuric acid;sulfur trioxide
SMILES O=S(=O)=O.OS(O)(=O)=O

Specifications

Density 1.925g/mL at 25°C, 1.90g/mL at 20°C
Residue after Ignition ≤0.01%
Vapor Pressure 2mmHg (25°C)
Quantity 1 L
Linear Formula H2SO4 · (SO3)x
UN Number 1831-
Beilstein 2037554
Formula Weight 98.08g/mol
Grade puriss. p.a.
Chemical Name or Material Sulfuric Acid, Fuming, 20% SO3
Hazard Statement P260-P280-P284-P305 + P351 + P338-P310
Precautionary Statement H314-H330-H335
RTECSNumber WS5605000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.