Learn More
Medchemexpress LLC Sulfuric acid, mono [(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester | 1192500-31-4 | 99.3% | 265.24 | 1 ML
SDP
Catalog No. 5000359549
Encompass_Preferred
Avibactam free acid, also known as NXL-104 free acid, is a potent covalent and reversible non-β-lactam β-lactamase inhibitor. It demonstrates strong inhibitory activity against β-lactamase TEM-1 and CTX-M-15, with IC50 values of 8 nM and 5 nM, respectively. This compound exhibits minimal antibacterial activity on its own but is effective in inhibiting Class A and C β-lactamases. It does not inhibit metallo β-lactamases or Acinetobacter OXA carbapenemases.
  • Broad-spectrum β-lactamase inhibition
  • Synergistic activity
  • In vivo efficacy
  • Research applications
Catalog No. 50-003-59549 Supplier Medchemexpress LLC Supplier No. HY1487910MM/1ML
Avibactam free acid, also known as NXL-104 free acid, is a potent covalent and reversible non-β-lactam β-lactamase inhibitor. It demonstrates strong inhibitory activity against β-lactamase TEM-1 and CTX-M-15, with IC50 values of 8 nM and 5 nM, respectively. This compound exhibits minimal antibacterial activity on its own but is effective in inhibiting Class A and C β-lactamases. It does not inhibit metallo β-lactamases or Acinetobacter OXA carbapenemases.
  • Broad-spectrum β-lactamase inhibition
  • Synergistic activity
  • In vivo efficacy
  • Research applications