Learn More
Medchemexpress LLC Sulfuric acid, mono(4-methylphenyl) ester | 3233-58-7 | 99.98% | 188.20 | 1 MG
SDP
Catalog No. 5000346436
Encompass_Preferred
p-Cresyl sulfate (p-Tolyl sulfate) is a uremic toxin that can cause renal damage and dysfunction. It shows antiproliferation activity, increases the protein expression of HIF-1α and VHL, decreases the protein expression of HIF-2α, induces epithelial-mesenchymal transition (EMT), and activates the JNK and p38 MAPK signaling pathways. For research use only.
  • Exhibits antiproliferation activity in a dose- and time-dependent manner
  • Increases protein expression of HIF-1α and VHL
  • Decreases protein expression of HIF-2α
  • Induces epithelial-mesenchymal transition (EMT)
  • Activates JNK and p38 MAPK signaling pathways
  • Inhibits lipogenesis and increases lipolysis in adipocytes
  • Induces osteoblast dysfunction
  • Induces ROS production in primary osteoblastic cells
Catalog No. 50-003-46436 Supplier Medchemexpress LLC Supplier No. HY11143110MM/1ML
p-Cresyl sulfate (p-Tolyl sulfate) is a uremic toxin that can cause renal damage and dysfunction. It shows antiproliferation activity, increases the protein expression of HIF-1α and VHL, decreases the protein expression of HIF-2α, induces epithelial-mesenchymal transition (EMT), and activates the JNK and p38 MAPK signaling pathways. For research use only.
  • Exhibits antiproliferation activity in a dose- and time-dependent manner
  • Increases protein expression of HIF-1α and VHL
  • Decreases protein expression of HIF-2α
  • Induces epithelial-mesenchymal transition (EMT)
  • Activates JNK and p38 MAPK signaling pathways
  • Inhibits lipogenesis and increases lipolysis in adipocytes
  • Induces osteoblast dysfunction
  • Induces ROS production in primary osteoblastic cells