Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sulfuric Acid Solution, 0.1M
SCI_ED

Catalog No. S25896 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
0.1M
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25896 500 mL 0.1M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25896

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging 0.1M
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sulfuric Acid Solution, 0.1M
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.