Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid Solution, Supelco

Catalog No. 11100250 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-250 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-250 Supplier MilliporeSigma Supelco Supplier No. 1.09984.0050
 Restricted Item
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Concentration 0.05 mol/L (0.1 N)
Quantity 1 Bottle
Molecular Weight (g/mol) 98.08
Percent Purity Unknown
Grade Ungraded
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.